Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Nidogen-2 ELISA Kit

Catalog No. EH338RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH338RBX5 5 x 96 Tests
EH338RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH338RBX5 Supplier Invitrogen™ Supplier No. EH338RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human Nidogen-2 quantitates human Nidogen-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nidogen-2.

Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis. Basement membranes, which are composed of type IV collagens, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14112
Assay Range 49-12000 pg/mL
Assay Sensitivity 49 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AW547149; entactin 2; entactin-2; LOW QUALITY PROTEIN: nidogen-2; Ly111; Nid2; NID-2; Nidogen; nidogen 2; nidogen 2 (osteonidogen); nidogen-2; nidogen-2; LOW QUALITY PROTEIN: nidogen-2; osteonidogenAW547149; entactin 2; entactin-2; LOW QUALITY PROTEIN: nidogen-2; Ly111; Nid2; NID-2; Nidogen; nidogen 2; nidogen 2 (osteonidogen); nidogen-2; nidogen-2; LOW QUALITY PROTEIN: nidogen-2; osteonidogen;
Gene ID (Entrez) 22795
Gene Symbol NID2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Nidogen 2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.