Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Nidogen Recombinant Protein, PeproTech®

Catalog No. 140175UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-017-5UG 5 μg
14-017-500UG 500 μg
14-017-25UG 25 μg
14-017-250UG 250 μg
14-017-1MG 1 mg
14-017-100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-017-5UG Supplier Gibco™ Supplier No. 140175UG
Add to Cart
Add to Cart

Recombinant Protein

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14543
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 4811
Molecular Weight (g/mol) 133.5 kDa
Name Human Nidogen
Purification Method Purified
Quantity 5 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Common Name Nidogen
Gene Symbol NID1
Biological Activity Determined by the ability of the immobilized protein to support the adhesion of HUVEC cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence LSRQELFPFG PGQGDLELED GDDFVSPALE LSGALRFYDR SDIDAVYVTT NGIIATSEPP AKESHPGLFP PTFGAVAPFL ADLDTTDGLG KVYYREDLSP SITQRAAECV HRGFPEISFQ PSSAVVVTWE SVAPYQGPSR DPDQKGKRNT FQAVLASSDS SSYAIFLYPE DGLQFHTTFS KKENNQVPAV VAFSQGSVGF LWKSNGAYNI FANDRESVEN LAKSSNSGQQ GVWVFEIGSP ATTNGVVPAD VILGTEDGAE YDDEDEDYDL ATTRLGLEDV GTTPFSYKAL RRGGADTYSV PSVLSPRRAA TERPLGPPTE RTRSFQLAVE TFHQQHPQVI DVDEVEETGV VFSYNTDSRQ TCANNRHQCS VHAECRDYAT GFCCSCVAGY TGNGRQCVAE GSPQRVNGKV KGRIFVGSSQ VPIVFENTDL HSYVVMNHGR SYTAISTIPE TVGYSLLPLA PVGGIIGWMF AVEQDGFKNG FSITGGEFTR QAEVTFVGHP GNLVIKQRFS GIDEHGHLTI DTELEGRVPQ IPFGSSVHIE PYTELYHYST SVITSSSTRE YTVTEPERDG ASPSRIYTYQ WRQTITFQEC VHDDSRPALP STQQLSVDSV FVLYNQEEKI LRYALSNSIG PVREGSPDAL QNPCYIGTHG CDTNAACRPG PRTQFTCECS IGFRGDGRTC YDIDECSEQP SVCGSHTICN NHPGTFRCEC VEGYQFSDEG TCVAVVDQRP INYCETGLHN CDIPQRAQCI YTGGSSYTCS CLPGFSGDGQ ACQDVDECQP SRCHPDAFCY NTPGSFTCQC KPGYQGDGFR CVPGEVEKTR CQHEREHILG AAGATDPQRP IPPGLFVPEC DAHGHYAPTQ CHGSTGYCWC VDRDGREVEG TRTRPGMTPP CLSTVAPPIH QGPAVPTAVI PLPPGTHLLF AQTGKIERLP LEGNTMRKTE AKAFLHVPAK VIIGLAFDCV DKMVYWTDIT EPSIGRASLH GGEPTTIIRQ DLGSPEGIAV DHLGRNIFWT DSNLDRIEVA KLDGTQRRVL FETDLVNPRG IVTDSVRGNL YWTDWNRDNP KIETSYMDGT NRRILVQDDL GLPNGLTFDA FSSQLCWVDA GTNRAECLNP SQPSRRKALE GLQYPFAVTS YGKNLYFTDW KMNSVVALDL AISKETDAFQ PHKQTRLYGI TTALSQCPQG HNYCSVNNGG CTHLCLATPG SRTCRCPDNT LGVDCIEQK
Content And Storage -20°C
Activity Determined by the ability of the immobilized protein to support the adhesion of HUVEC cells.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.