Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NIFK (aa 181-261) Control Fragment Recombinant Protein

Catalog No. RP104083
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104083 Supplier Invitrogen™ Supplier No. RP104083
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83283. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NIFK, also known as nucleolar protein interacting with the forkhead homolog protein, is a gene associated with several cancer types and is considered a potential prognostic biomarker. This gene functions within the nucleolus and plays a critical role in cellular proliferation, metastasis, and invasiveness, particularly noted in hepatocellular carcinoma (HCC) and colorectal cancer. NIFK expression is regulated via several pathways, including the MYC proto-oncogene pathway, and its silencing can lead to decreased fatty acid synthesis in colorectal cancer cells. Furthermore, it's involved in the regulation of innate immunity through pathways such as NF-kB in specific cancer cells. Due to its involvement in cancer progression, NIFK is being explored as a therapeutic target and holds promise for advancing precision medicine strategies across various cancer treatments.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BYG3
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 84365
Name Human NIFK (aa 181-261) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI852665; C130020J04Rik; hNIFK; Ki67ip; MKI67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; MKI67IP; mNIFK; NIFK; NOPP34; nucleolar phosphoprotein Nopp34; nucleolar protein interacting with the FHA domain of MKI67; nucleolar protein interacting with the FHA domain of pKI-67; spermatogenesis-related protein (Srp)
Common Name NIFK
Gene Symbol Nifk
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IDYDFPSLILQKTESISKTNRQTSTKGQVLRKKKKKVSGTLDTPEKTVDSQGPTPVCTPTFLERRKSQVAELNDDDKDDEI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.