Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Ninjurin 2 (aa 84-110) Control Fragment Recombinant Protein

Catalog No. RP109124
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109124 Supplier Invitrogen™ Supplier No. RP109124
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140118. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Ninjurin 2 (NINJ2) is a gene located on chromosome 12p13.2, encoding a member of the ninjurin family, which consists of cell surface proteins involved in promoting cell-cell adhesion and nerve regeneration. NINJ2 plays a pivotal role in neural repair processes, particularly in the peripheral nervous system, where it contributes to axonal growth and the formation of new neural connections following injury. This protein is expressed in various tissues, including the nervous system and endothelial cells, and is implicated in modulating inflammatory responses and cell-cell interactions. In addition to its role in the nervous system, NINJ2 is associated with vascular biology and immune cell migration, influencing processes such as angiogenesis and leukocyte transmigration. Altered expression or mutations in NINJ2 have been studied in the context of neurodegenerative diseases and stroke, where it may affect recovery and repair mechanisms. Research continues to explore NINJ2's potential as a therapeutic target for enhancing nerve regeneration and addressing vascular and inflammatory conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZG7
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 4815
Name Human Ninjurin 2 (aa 84-110) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias FLJ56349; Nerve injury-induced protein 2; NINJ2; ninjurin 2; ninjurin-2
Common Name Ninjurin 2
Gene Symbol NINJ2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LVVIARLNLNEVEKQWRLNQLNNAATI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.