Learn More
STEMCELL Technologies EasySep™ Human NK Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0236166
Encompass
The EasySep™ Human NK Cell Enrichment Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-NK cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human NK Cell Isolation Kit (17955) (/products/easysep-human-nk-cell-isolation-kit.html) which isolates cells in just 8 minutes.
Catalog No. NC0236166 Supplier STEMCELL Technologies Supplier No. 19055
The EasySep™ Human NK Cell Enrichment Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-NK cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human NK Cell Isolation Kit (17955) (/products/easysep-human-nk-cell-isolation-kit.html) which isolates cells in just 8 minutes.