Learn More
ACROBiosystems Human NKG2A CD94 100ug
Catalog No. 502534398
Encompass_Preferred
Human NKG2ACD94 Protein His Tag (NC4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) Q13241-1 (CD94)).
Catalog No. 50-253-4398 Supplier ACROBiosystems Supplier No. NC4H52H3100UG
Human NKG2ACD94 Protein His Tag (NC4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) Q13241-1 (CD94)).