Learn More
Sino Biological Recombinant Human NKG2A Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 13905H01H100
Encompass_Preferred
A DNA sequence encoding the human KLRC1 (NP_002250.1) (Pro94-Leu233) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-224-7218