Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human NKG2D Fc Recombinant Protein, PeproTech®
GREENER_CHOICE

Catalog No. 5000331921
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-003-31921 100 μg
31-051-10UG 10 μg
31-051-1MG 1 mg
50-003-31935 250 μg
50-003-31936 500 μg
31-051-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 50-003-31921 Supplier Gibco™ Supplier No. 31051100UG
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human NKG2D Fc is an 84.8 kDa glycosylated, disulfide-linked homodimer whose 374 amino acid monomer consists of the 139 amino acid N-terminal portion of the extracellular domain fused to the 233-amino-acid residue C-terminal Fc region of human IgG by two glycine residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P26718
For Use With (Application) Binding Assay, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 22914
Molecular Weight (g/mol) 42.4 kDa
Name Human NKG2D Fc
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D
Common Name NKG2D (CD314)
Gene Symbol Klrk1
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence MDPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGKGGFLNSL FNQEVQIPLT ESYCGPCPKN WICYKNNCYQ FFDESKNWYE SQASCMSQNA SLLKVYSKED QDLLKLVKSY HWMGLVHIPT NGSWQWEDGS ILSPNLLTII EMQKGDCALY ASSFKGYIEN CSTPNTYICM QRTV
Activity Determined by its ability to bind human MICA/Fc Chimera in a functional ELISA.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.