Learn More
Raybiotech Inc Human NKp44 ELISA Kit
Supplier:  Raybiotech Inc ELHNKP441
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 13 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human NKp44.
Catalog No. 50-194-9583