Learn More
Sino Biological Anti-Neuroligin 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11617RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Neuroligin 1 / NLGN1 (rh Neuroligin 1 / NLGN1; Catalog#11617-H08H; NP_055747.1; Met 1-Ser 677). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4662