Learn More
Sino Biological Anti-Neuroligin 3/NLGN3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11160T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Neuroligin 3 / NLGN3 (rh Neuroligin 3 / NLGN3; Catalog#11160-H08H; Q9NZ94-2; Met1-Ser689). Neuroligin 3 / NLGN3 specific IgG was purified by Human Neuroligin 3 / NLGN3 affinity chromatography.
Catalog No. 50-241-0754