Learn More
Sino Biological Anti-Neuroligin 3/NLGN3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201759T02100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Neuroligin 3 / NLGN3 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2083