Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NLRC4 (aa 756-889) Control Fragment Recombinant Protein

Catalog No. RP89236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89236 Supplier Invitrogen™ Supplier No. RP89236
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NLRC4 encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NPP4
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 58484
Name Human NLRC4 (aa 756-889) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 9530011P19Rik; AIFEC; CARD, LRR, and NACHT-containing protein; Card12; caspase recruitment domain family, member 12; caspase recruitment domain-containing protein 12; CLAN; Clan protein; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; ice protease-activating factor; ICE-protease activating factor; ipaf; NLR family CARD domain containing 4; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NLRC4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4; UNQ6189/PRO20215
Common Name NLRC4
Gene Symbol Nlrc4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TDSLGNLKNLTKLIMDNIKMNEEDAIKLAEGLKNLKKMCLFHLTHLSDIGEGMDYIVKSLSSEPCDLEEIQLVSCCLSANAVKILAQNLHNLVKLSILDLSENYLEKDGNEALHELIDRMNVLEQLTALMLPWG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.