Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NLRX1 Synthetic Peptide

Catalog No. PEP1132 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1132 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1132 Supplier Invitrogen™ Supplier No. PEP1132
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 14 amino acids near the amino terminus of human NOD5. PEP-1132 can be used as a blocking peptide with polyclonal antibody PA5-21018.

This gene encodes a member of the NLR family. Alternative splicing has been observed at this gene locus and two transcript variants, encoding distinct isoforms, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 79671
Species Human
Purification Method purified
Accession Number Q86UT6
Common Name NLRX1
Concentration 1 mg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias BC034204; Caterpiller protein 11.3; CLR11.3; DLNB26; FLJ21478; MGC131937; MGC21025; NLR family member X1; NLRX1; NOD26; NOD5; NOD9; NOD-like receptor X1; nucleotide-binding domain and leucine-rich repeat containing; nucleotide-binding oligomerization domain protein 26; Nucleotide-binding oligomerization domain protein 5; nucleotide-binding oligomerization domain protein 9; nucleotide-binding oligomerization domain, leucine rich repeat containing X1; OTTHUMP00000205768; OTTHUMP00000205769; RGD1311293
Gene Symbol NLRX1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.