Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NMD3 Full-length ORF (NP_057022.2, 1 a.a. - 503 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7100341 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-954-277 25 μg
89-954-276 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-954-277 Supplier Abnova™ Supplier No. H00051068P01L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for AP, Array, ELISA, WB-Re

Ribosomes are composed of 60S and 40S subunits that are assembled in the nucleolus and exported to the cytoplasm through nuclear pore complexes in the nuclear envelope. NMD3 is an adaptor for 60S subunit export via the CRM1 (XPO1; MIM 602559) pathway (Trotta et al., 2003 [PubMed 12773398]).[supplied by OMIM]

Sequence: MEYMAESTDRSPGHILCCECGVPISPNPANICVACLRSKVDISQGIPKQVSISFCKQCQRYFQPPGTWIQCALESRELLALCLKKIKAPLSKVRLVDAGFVWTEPHSKRLKVKLTIQKEVMNGAILQQVFVVDYVVQSQMCGDCHRVEAKDFWKAVIQVRQKTLHKKTFYYLEQLILKYGMHQNTLRIKEIHDGLDFYYSSKQHAQKMVEFLQCTVPCRYKASQRLISQDIHSNTYNYKSTFSVEIVPICKDNVVCLSPKLAQSLGNMNQICVCIRVTSAIHLIDPNTLQVADIDGSTFWSHPFNSLCHPKQLEEFIVMECSIVQDIKRAAGAGMISKKHTLGEVWVQKTSEMNTDKQYFCRTHLGHLLNPGDLVLGFDLANCNLNDEHVNKMNSDRVPDVVLIKKSYDRTKRQRRRNWKLKELARERENMDTDDERQYQDFLEDLEEDEAIRKNVNIYRDSAIPVESDTDDEGAPRISLAEMLEDLHISQDATGEEGASMLT

Specifications

Accession Number NP_057022.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51068
Molecular Weight (g/mol) 84kDa
Name NMD3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MEYMAESTDRSPGHILCCECGVPISPNPANICVACLRSKVDISQGIPKQVSISFCKQCQRYFQPPGTWIQCALESRELLALCLKKIKAPLSKVRLVDAGFVWTEPHSKRLKVKLTIQKEVMNGAILQQVFVVDYVVQSQMCGDCHRVEAKDFWKAVIQVRQKTLHKKTFYYLEQLILKYGMHQNTLRIKEIHDGLDFYYSSKQHAQKMVEFLQCTVPCRYKASQRLISQDIHSNTYNYKSTFSVEIVPICKDNVVCLSPKLAQSLGNMNQICVCIRVTSAIHLIDPNTLQVADIDGSTFWSHPFNSLCHPKQLEEFIVMECSIVQDIKRAAGAGMISKKHTLGEVWVQKTSEMNTDKQYFCRTHLGHLLNPGDLVLGFDLANCNLNDEHVNKMNSDRVPDVVLIKKSYDRTKRQRRRNWKLKELARERENMDTDDERQYQDFLEDLEEDEAIRKNVNIYRDSAIPVESDTDDEGAPRISLAEMLEDLHISQDATGEEGASMLT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CGI-07/FLJ21053
Common Name NMD3
Gene Symbol NMD3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.