Learn More
Sino Biological Anti-NNT Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104378T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NNT, and purified by antigen affinity chromatography.
Catalog No. 50-241-0579