Learn More
Sino Biological Recombinant Human Noggin/NOG Protein (hFc Tag) 1 mg
Supplier:  Sino Biological 10267H02H1
Encompass_Preferred
A DNA sequence encoding the human Noggin precursor (NP_005441.1) (Met 1-Cys 232) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7011