Learn More
Raybiotech Inc Human Nogo-B/RTN 4B ELISA Kit
Supplier:  Raybiotech Inc ELHNOGOB1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 13 pg/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Nogo-B.
Catalog No. 50-194-9586