Learn More
Sciencell Research Laboratories Human Non-Pigmented Ciliary Epithelial Cells Kit
Catalog No. NC3874091
Encompass
solated from human ciliary body. HNPCEpiC are cryopreserved at P0 and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.Kit includes:Cells (Catalog #6580), Specialty Medium (Catalog #4101), DPBS (Catalog #0303), Trypsin/EDTA (Catalog #0183), Trypsin Neutralizing Solution (Catalog #0113), PLL (Catalog #0413)
Catalog No. NC3874091 Supplier Sciencell Research Laboratories Supplier No. 6580KIT
solated from human ciliary body. HNPCEpiC are cryopreserved at P0 and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.Kit includes:Cells (Catalog #6580), Specialty Medium (Catalog #4101), DPBS (Catalog #0303), Trypsin/EDTA (Catalog #0183), Trypsin Neutralizing Solution (Catalog #0113), PLL (Catalog #0413)