Learn More
Sciencell Research Laboratories Human Non-Pigmented Ciliary Epithelial Cells only
Catalog No. NC3874086
Encompass
Isolated from human ciliary body. HNPCEpiC are cryopreserved at P0 and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.
Catalog No. NC3874086 Supplier Sciencell Research Laboratories Supplier No. 6580
Isolated from human ciliary body. HNPCEpiC are cryopreserved at P0 and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.