Learn More
Amsbio LLC Human Normal Liver FFPE Tissue Block
Catalog No. 502041370
Encompass
Human FFPE tissue blocks are processed from diseased tissues or clinically-unremarkable donor normal human tissue samples. Tissue is prepared by a certified medical pathologist, fixed in 10% neutral formalin for 24 hours and embedded in IHC-grade paraffin. The target FFPE specimen size is 0.5 x 0.5 x 0.5 cm, but it can vary significantly depending on the organ.
Catalog No. 50-204-1370 Supplier Amsbio LLC Supplier No. TPH0300B
Human FFPE tissue blocks are processed from diseased tissues or clinically-unremarkable donor normal human tissue samples. Tissue is prepared by a certified medical pathologist, fixed in 10% neutral formalin for 24 hours and embedded in IHC-grade paraffin. The target FFPE specimen size is 0.5 x 0.5 x 0.5 cm, but it can vary significantly depending on the organ.