Learn More
Biomatik Corporation Human NOS2/iNOS (Nitric oxide synthase, inducible) ELISA Kit
Supplier:  Biomatik Corporation EKF58509
Encompass_Preferred
Human NOS2/iNOS (Nitric oxide synthase, inducible) ELISA Kit for detecting NOS2; iNOS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35228. Target Synonyms: NOS2; iNOS; NOS2A; HEP-NOS; I-NOS; Hepatocyte NOS; Peptidyl-cysteine S-nitrosylase NOS2; Inducible NO synthase; Inducible NOS; INOS; NOS; NOS; type II; NOS type II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9174