Learn More
Raybiotech Inc Human Notch-1 ELISA Kit
Supplier:  Raybiotech Inc ELHNOTCH11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 20 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The sandwich ELISA antibody pair detects Human NOTCH-1.
Catalog No. 50-194-9589