Learn More
BIOHIPPO Human NOTCH2NL Microsample
SDP
Catalog No. 5000315380
Encompass
Human NOTCH2NL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NOTCH2NL in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NOTCH2NL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NOTCH2NL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NOTCH2NL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15380 Supplier BIOHIPPO Supplier No. BHE15211027
Human NOTCH2NL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NOTCH2NL in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NOTCH2NL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NOTCH2NL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NOTCH2NL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color