Learn More
Sino Biological Anti-NOV/CCN3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10264MM0920
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NOV/CCN3 (rh NOV/CCN3; Catalog#10264-H08B; NP_002505.1; Met1-Met357). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2377