Learn More
Sino Biological Recombinant Anti-NOV/CCN3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10264R13020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCN3 / NOV / IGFBP9 (rh CCN3 / NOV / IGFBP9; Catalog#10264-H08B; NP_002505.1; Met 1-Met 357).
Catalog No. 50-241-0316