Learn More
Sino Biological Recombinant Anti-Niemann Pick C2/NPC2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13341R137100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human NPC2 (rh NPC2; Catalog#13341-H08H; NP_006423.1; Met1-Leu151).
Catalog No. 50-255-4254