Learn More
Sino Biological Anti-Niemann Pick C2/NPC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13341RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Niemann-Pick disease type C2 / NPC2 (rh Niemann-Pick disease type C2 / NPC2; Catalog#13341-H08H; NP_006423.1; Met1-Leu151). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4253