Learn More
Sino Biological Recombinant Human Nephrin Protein (ECD, His Tag)
Supplier:  Sino Biological 17757H08H1
Encompass_Preferred
A DNA sequence encoding the Human NPHS1 (O60500) (Met1-Ser1055) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2100