Learn More
Sino Biological Human NPHS2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 102168T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Podocin / NPHS2, and purified by antigen affinity chromatography.
Catalog No. 50-237-3255