Learn More
BIOHIPPO Human NPNT Microsample
SDP
Catalog No. 5000314717
Encompass
Human NPNT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NPNT in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NPNT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NPNT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NPNT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14717 Supplier BIOHIPPO Supplier No. BHE15211790
Human NPNT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NPNT in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NPNT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NPNT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NPNT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color