Learn More
Sino Biological Anti-NPTN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15881RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NPTN / SDFR1 (rh NPTN; Catalog#15881-H08H; NP_059429.1; Met1-Leu221). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4079