Learn More
Sino Biological Anti-NPTN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15881T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NPTN / SDFR1 (rh NPTN / SDFR1; Catalog#15881-H08H; NP_059429.1; Met1-Leu221). NPTN / SDFR1 specific IgG was purified by Human NPTN / SDFR1 affinity chromatography.
Catalog No. 50-255-4078