Learn More
Sino Biological Recombinant Human Farnesoid X receptor/FXR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 17624H07E50
Encompass_Preferred
A DNA sequence encoding the Human NR1H4 (Q96RI1-1) (Thr247−Gln476) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-241-1183