Learn More
Sino Biological Human NR3C1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 201121T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human NR3C1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-0718