Learn More
Sino Biological Human NR3C1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 12044H08E100
Encompass_Preferred
A DNA sequence encoding the Human NR3C1 (P04150) (Pro417-Gly506) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3578