Learn More
Sino Biological Anti-NRG1 Alpha Antibody
Supplier:  Sino Biological 13499T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NRG1 (rh NRG1; Catalog#13499-H08H; Q02297-1; Ser20-Lys241). NRG1 specific IgG was purified by Human NRG1 affinity chromatography.
Catalog No. 50-255-4227