Learn More
Sino Biological Recombinant Human Neuregulin-1/NRG1 Protein (ECD), HPLC-verified
Supplier:  Sino Biological 11609HNAE100
Encompass_Preferred
A DNA sequence encoding the Human NRG1 (Q02297-6) (Thr176-Lys246) was expressed with an N-terminal Met.
Catalog No. 50-255-4681