Learn More
Sino Biological Recombinant Human Neuregulin-1/NRG1 Protein (ECD), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11609HNCHB20
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Ser2-Lys246) of human NRG1 isoform beta1 (Q02297-6) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4678