Learn More
Sino Biological Anti-Neuregulin-3/NRG3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 16071MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NRG-3 / Neuregulin-3 (rh NRG-3 / Neuregulin-3; Catalog#16071-H08H; ADK90021.1; Glu282-Pro342). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4064