Learn More
Sino Biological Human NRG3 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 16071H08H1
Encompass_Preferred
A DNA sequence encoding the human NRG3 (ADK90021.1) (Glu282-Pro342) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3401