Learn More
Sino Biological Recombinant Anti-Neuregulin-4/NRG4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12183R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human NRG4 (rh NRG4; Catalog#12183-HNCE; Q8WWG1; Pro2-Phe62).
Catalog No. 50-255-4422