Learn More
Sino Biological Anti-Neuregulin-4/NRG4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12183T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NRG4 (rh NRG4; Catalog#12183-HNCE; Q8WWG1; Pro2-Phe62). NRG4 specific IgG was purified by Human NRG4 affinity chromatography.
Catalog No. 50-255-4421