Learn More
Sino Biological Human NRP1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10011H08H21
Encompass_Preferred
A DNA sequence encoding the human NRP1 (NP_001019799.2) (Met1-Lys644) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3865