Learn More
Sino Biological Anti-Neuropilin-2/NRP2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10695MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Neurophilin-2 extracellular domain (rhNRP2; Catalog#10695-H08H; Met1-Tyr855; NP_003863.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8485