Learn More
BIOHIPPO Human NRP2 Microsample
SDP
Catalog No. 5000315329
Encompass
Human NRP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NRP2 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 33 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NRP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NRP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NRP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15329 Supplier BIOHIPPO Supplier No. BHE15211880
Human NRP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NRP2 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 33 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NRP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NRP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NRP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color