Learn More
Sino Biological Anti-Neuropilin-2/NRP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10695RP03100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NRP2 extracellular domain (rhNRP2; Catalog#10695-H08H; Met1-Tyr855; NP_003863.2). NRP2 specific IgG was purified by human NRP2 affinity chromatography.
Catalog No. 50-241-0968