Learn More
Sino Biological Anti-Neurexin-3 beta/NRXN3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11843T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NRXN3 (rh NRXN3; Catalog#11843-H08H; NP_620426.2; Met1-Thr357). NRXN3 specific IgG was purified by Human NRXN3 affinity chromatography.
Catalog No. 50-255-4523