Learn More
Sino Biological Human NRXN3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11843RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NRXN3 / Neurexin 3 (rh NRXN3 / Neurexin 3; Catalog#11843-H08H; NP_620426.2; Met1-Thr357). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-0829