Learn More
Sino Biological Anti-NSL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107134T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NSL1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8760